Learn More
AVANTOR MASTERFLEX (VWR LLC) Gauge Guard Isolator Sanitary Gasket, Platinum-cured Silicone, 3/4"" Clamp Size
Catalog No. NC1805877
Encompass
Simple accessories to protect your delicate pressure gauge. Choose from platinum-cured silicone, FKM, EPDM or PTFE materials.
Catalog No. NC1805877 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3056502
Simple accessories to protect your delicate pressure gauge. Choose from platinum-cured silicone, FKM, EPDM or PTFE materials.