Learn More
AVANTOR MASTERFLEX (VWR LLC) Gauge Guard Isolator Sanitary Gasket, Platinum-cured Silicone, 1-1/2"" Clamp Size
Catalog No. NC1457592
Encompass
Gauge Guard Isolator Sanitary Gasket, Platinum-cured Silicone, 1-1/2" Clamp Size, Gaskets feature sanitary seal edges and a full membrane made of the same material as the sealing gasket edge. No more worry about particulates and film building up on your pressure membrane! Available in platinum-cured silicone, FKM, EPDM and PTFE.
Catalog No. NC1457592 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3056506
Gauge Guard Isolator Sanitary Gasket, Platinum-cured Silicone, 1-1/2" Clamp Size, Gaskets feature sanitary seal edges and a full membrane made of the same material as the sealing gasket edge. No more worry about particulates and film building up on your pressure membrane! Available in platinum-cured silicone, FKM, EPDM and PTFE.