Learn More
Ace Glass, Inc. Gauge, McLeod, 10mm Hg, scale only
SDP
Catalog No. 50976242
Encompass_Preferred
0-10mm Hg Scale only for McLeod vacuum gauge. McLeod vacuum gauge is accurate to within +/- 3%. Mercury NOT supplied.
Catalog No. 50-976-242 Supplier Ace Glass, Inc. Supplier No. 872634
0-10mm Hg Scale only for McLeod vacuum gauge. McLeod vacuum gauge is accurate to within +/- 3%. Mercury NOT supplied.