Learn More
Ace Glass, Inc. Gauge, McLeod, 10mm Hg, volumetric leg only
SDP
Catalog No. 50976250
Encompass_Preferred
A 10MM tilting gauge which maintains a constant overflow head of mercury and maintains capillary depression of the meniscus.
Catalog No. 50-976-250 Supplier Ace Glass, Inc. Supplier No. 872843
A 10MM tilting gauge which maintains a constant overflow head of mercury and maintains capillary depression of the meniscus.