Learn More
Ace Glass, Inc. 0-15mm Hg Scale only for McLeod vacuum gauge
SDP
Catalog No. 50976243
Encompass_Preferred
0-15mm Hg Scale only for McLeod vacuum gauge. McLeod vacuum gauge is accurate to within +/- 3%. Mercury NOT supplied.
Catalog No. 50-976-243 Supplier Ace Glass, Inc. Supplier No. 872636
0-15mm Hg Scale only for McLeod vacuum gauge. McLeod vacuum gauge is accurate to within +/- 3%. Mercury NOT supplied.