Learn More
Ace Glass, Inc. Gauge, McLeod, 15mm Hg, volumetric leg only
SDP
Catalog No. 50979823
Encompass_Preferred
A 15MM tilting gauge which maintains a constant overflow head of mercury and maintains capillary depression of the meniscus.
Catalog No. 50-979-823 Supplier Ace Glass, Inc. Supplier No. 872845
A 15MM tilting gauge which maintains a constant overflow head of mercury and maintains capillary depression of the meniscus.