Learn More
Ace Glass, Inc. 0-1mm Hg Scale only for McLeod vacuum gauge
SDP
Catalog No. 50976241
Encompass_Preferred
0-1mm Hg Scale only for McLeod vacuum gauge. McLeod vacuum gauge is accurate to within +/- 3%. Mercury NOT supplied.
Catalog No. 50-976-241 Supplier Ace Glass, Inc. Supplier No. 872630
0-1mm Hg Scale only for McLeod vacuum gauge. McLeod vacuum gauge is accurate to within +/- 3%. Mercury NOT supplied.