Learn More
Ace Glass, Inc. Gauge, McLeod, 20/65 joint only
SDP
Catalog No. 50976240
Encompass_Preferred
20/65 standard taper joint for McLeod vacuum gauge. McLeod vacuum gauge is accurate to within +/- 3% and measures pressures between 0-15mm Hg. Mercury NOT supplied.
Catalog No. 50-976-240 Supplier Ace Glass, Inc. Supplier No. 872624
20/65 standard taper joint for McLeod vacuum gauge. McLeod vacuum gauge is accurate to within +/- 3% and measures pressures between 0-15mm Hg. Mercury NOT supplied.