Learn More
Ace Glass, Inc. Gauge, McLeod, 5mm Hg, glass body and scale
SDP
Catalog No. 50976247
Encompass_Preferred
A 5MM tilting gauge which maintains a constant overflow head of mercury and maintains capillary depression of the meniscus.
Catalog No. 50-976-247 Supplier Ace Glass, Inc. Supplier No. 872813
A 5MM tilting gauge which maintains a constant overflow head of mercury and maintains capillary depression of the meniscus.