Learn More
Ace Glass, Inc. 0-5mm Hg Scale only for McLeod vacuum gauge
SDP
Catalog No. 50979820
Encompass_Preferred
0-5mm Hg Scale only for McLeod vacuum gauge. McLeod vacuum gauge is accurate to within +/- 3%. Mercury NOT supplied.
Catalog No. 50-979-820 Supplier Ace Glass, Inc. Supplier No. 872632
0-5mm Hg Scale only for McLeod vacuum gauge. McLeod vacuum gauge is accurate to within +/- 3%. Mercury NOT supplied.