Learn More
Cole-Parmer Cole-Parmer Pressure Gauge, Viton diaphragm seal, 1/4" NPT(F); 0 to 15 psi
Catalog No. NC1212737 Shop All Cole-Parmer Products
Encompass_Preferred
These gauges are molded from a harder blend of polymers, making them more resistant to cracking or breaking during installation and use. Gauges feature brass Bourdon tube and internals; diaphragm fill is glycerin. White PVDF gauges feature a PTFE diaphragm seal; black polypropylene (PP) gauges feature a Viton diaphragm seal. All gauge casings are dry but can be filled.
Catalog No. NC1212737 Supplier Cole-Parmer Supplier No. 6800025
These gauges are molded from a harder blend of polymers, making them more resistant to cracking or breaking during installation and use. Gauges feature brass Bourdon tube and internals; diaphragm fill is glycerin. White PVDF gauges feature a PTFE diaphragm seal; black polypropylene (PP) gauges feature a Viton diaphragm seal. All gauge casings are dry but can be filled.