Learn More
Cole-Parmer Ashcroft 1009SWL2.5 0 to 100 psi Filled High-Accuracy Gauge 2 1/2" Dial Bottom Connection
Catalog No. 50992669 Shop All Cole-Parmer Products
Encompass_Preferred
Gauges feature plastic (polycarbonate) lens, and 316 stainless steel socket, tube, and case. All connections are 1/4" NPT(M).
Catalog No. 50-992-669 Supplier Cole-Parmer Supplier No. 6802204
Gauges feature plastic (polycarbonate) lens, and 316 stainless steel socket, tube, and case. All connections are 1/4" NPT(M).