Learn More
Cole-Parmer Ashcroft 1005H2.0 Utility Gauge, 0 to 100 psi, 2" Dial, Bottom Connection
Supplier:  Cole-Parmer 6800304
Encompass_Preferred
Gauges include brass socket, soft-soldered phosphor bronze Bourdon tube, painted steel case, and polycarbonate lens.
Catalog No. 50-992-661
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov