Learn More
Cole-Parmer Ashcroft 30,052.5 Commercial Gauge with Bottom Connection, 100 psi, 2-1/2" Dial
Catalog No. 50992657 Shop All Cole-Parmer Products
Encompass_Preferred
Use these glycerin-filled gauges for easier reading when high-frequency pressure fluctuations or vibrations are present. Gauges construction includes brass socket, phosphor bronze tube, 304 stainless steel case, and plastic lens. Bottom and back connections are 1/4" NPT(M).
Catalog No. 50-992-657 Supplier Cole-Parmer Supplier No. 0738080
Use these glycerin-filled gauges for easier reading when high-frequency pressure fluctuations or vibrations are present. Gauges construction includes brass socket, phosphor bronze tube, 304 stainless steel case, and plastic lens. Bottom and back connections are 1/4" NPT(M).