Learn More
Cole-Parmer Ashcroft 1005PH1.5 Utility Gauge, 0 to 100 psi, 1 1/2" Dial, Back Connection
Catalog No. 50992660 Shop All Cole-Parmer Products
Encompass_Preferred
Gauges include brass socket, soft-soldered phosphor bronze Bourdon tube, painted steel case, and polycarbonate lens.
Catalog No. 50-992-660 Supplier Cole-Parmer Supplier No. 6800204
Gauges include brass socket, soft-soldered phosphor bronze Bourdon tube, painted steel case, and polycarbonate lens.