Learn More
Cole-Parmer Ashcroft 1008SL2.5 0 to 160 psi and 0 to 1100 kPa Dual-Scale Gauge, Bottom Connection
Catalog No. 50992665 Shop All Cole-Parmer Products
Encompass_Preferred
Gauge construction includes acrylic lens and 316 stainless steel socket, Boudon tube, and case. Welded sealed joints protect against corrosion.
Catalog No. 50-992-665 Supplier Cole-Parmer Supplier No. 6800706
Gauge construction includes acrylic lens and 316 stainless steel socket, Boudon tube, and case. Welded sealed joints protect against corrosion.