Learn More
Gauge Pressure Demonstration
SCI_ED
Catalog No. S96168
Change view
Click to view available options
Gauge Pressure Demonstration
Catalog No.
S96168 Gauge Pressure Demonstration
1 options
Catalog No. S96168 Supplier Transparent Devices Supplier No. PRESS02
Includes
Instructions and experiments
Requires
Bell Jar and Vacuum Pump (S96167).
Measure atmospheric pressure as well as clarify the distinction between gauge pressure and absolute pressure
  • Durable plastic construction for years of classroom use
Specifications
Product Type Gauge Pressure Demonstration
For Use With (Application) Measure atmospheric pressure as well as clarify the distinction between gauge pressure and absolute pressure
Includes Instructions and experiments