Learn More
Chemglass Life Sciences Gauge, Pressure, Stainless Steel Connection and Case, 0-15 PSI, 1/4" M NPT, 2" Dia
SDP
Catalog No. 50143585
Encompass_Preferred
Gauge, Pressure, S.S. Connection and Case, 0-15 PSI, 1/4" M NPT, 2" Dia. PTFE stopper is similar to CG-3002-L but with the addition of one female NPT port in the center for connection of compression fittings or pressure/vacuum gauges. PTFE stopper has an extraction nut permitting easy removal of the stopper from the glass outer joint.
Catalog No. 50-143-585 Supplier Chemglass Life Sciences Supplier No. CG3002K80
Gauge, Pressure, S.S. Connection and Case, 0-15 PSI, 1/4" M NPT, 2" Dia. PTFE stopper is similar to CG-3002-L but with the addition of one female NPT port in the center for connection of compression fittings or pressure/vacuum gauges. PTFE stopper has an extraction nut permitting easy removal of the stopper from the glass outer joint.