Learn More
Cole-Parmer Ashcroft 1259SD4.5 4.5" Industrial Process Gauge 0 to 30 psi
Catalog No. 50992704 Shop All Cole-Parmer Products
Encompass_Preferred
Industries for these rugged process gauges include refineries, petrochemical, chemical, wastewater, and pulp and paper. Bourdon tube is all welded for safety and longer life. Range is vacuum to 20,000 psi.
Heavy- duty gauges are available filled and unfilled; liquid fill is glycerin. Gauge ring features cam lock and fiberglass reinforced thermoplastic; movement is 300 series stainless steel. Gauges window is glass. Proof pressure of these industrial gauges is 130% of full scale.
Catalog No. 50-992-704 Supplier Cole-Parmer Supplier No. 6833706
Industries for these rugged process gauges include refineries, petrochemical, chemical, wastewater, and pulp and paper. Bourdon tube is all welded for safety and longer life. Range is vacuum to 20,000 psi.
Heavy- duty gauges are available filled and unfilled; liquid fill is glycerin. Gauge ring features cam lock and fiberglass reinforced thermoplastic; movement is 300 series stainless steel. Gauges window is glass. Proof pressure of these industrial gauges is 130% of full scale.