Learn More
Cole-Parmer PVDF Pressure Gauge, PTFE diaphragm seal, 1/4" NPT(F); 0 to 100 psi
Catalog No. NC2175809 Shop All Cole-Parmer Products
Encompass
These gauges are molded from a harder blend of polymers, making them more resistant to cracking or breaking during installation and use. Gauges feature brass Bourdon tube and internals; diaphragm fill is glycerin. White PVDF gauges feature a PTFE diaphragm seal; black polypropylene (PP) gauges feature a Viton diaphragm seal. All gauge casings are dry but can be filled.
This item is not returnable. View return policy
Catalog No. NC2175809 Supplier Cole-Parmer Supplier No. 6800007
These gauges are molded from a harder blend of polymers, making them more resistant to cracking or breaking during installation and use. Gauges feature brass Bourdon tube and internals; diaphragm fill is glycerin. White PVDF gauges feature a PTFE diaphragm seal; black polypropylene (PP) gauges feature a Viton diaphragm seal. All gauge casings are dry but can be filled.