Learn More
Chemglass Life Sciences Gauge Shelf, Airfree, Schlenk
SDP
Catalog No. AF030501
Encompass_Preferred
Gauge Shelf, Airfree, Schlenk. Shelf is constructed of plexiglass with stainless steel support rods and is supplied complete with two bossheads for attachment to the uprights of the AF-0300-02 cart assembly. Center to center on support rods is 136mm.
Catalog No. AF030501 Supplier Chemglass Life Sciences Supplier No. AF030501
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Gauge Shelf, Airfree, Schlenk. Shelf is constructed of plexiglass with stainless steel support rods and is supplied complete with two bossheads for attachment to the uprights of the AF-0300-02 cart assembly. Center to center on support rods is 136mm.