Learn More
Cole-Parmer Wika 332.54 4 4" Test Gauge, 0 to 100 psi, 316 SS
Supplier:  Cole-Parmer 6804308
Encompass_Preferred
Gauge construction includes a 316 stainless steel (SS) socket; a 316 SS Bourdon tube for the 4" dial gauges or a copper alloy tube for the 6" dial gauges; a 316 SS fitting; a 304 SS case; and a glass lens. All gauges have a 1/4" NPT(M) process connection. The 4" dial gauges include a zippered, soft nylon carrying case.
Catalog No. 50-993-686