Learn More
Vacuum Research Corp GAUGE TUBE PIRANI 2000 MTORR
SDP
Catalog No. 50304086
Encompass
Pirani gauge tube, 1-2000 mTorr, 1/8 NPT
Catalog No. 50-304-086 Supplier Vacuum Research Corp Supplier No. 912001
Pirani gauge tube, 1-2000 mTorr, 1/8 NPT