Learn More
Vacuum Research Corp GAUGE TUBE THERMOCOUPLE DV-6
SDP
Catalog No. 50304108
Encompass
VRC-6M Thermocouple gauge tube, interchangeable with Hastings DV-6M and DV-6R, 1/8 NPT
Catalog No. 50-304-108 Supplier Vacuum Research Corp Supplier No. 912086
VRC-6M Thermocouple gauge tube, interchangeable with Hastings DV-6M and DV-6R, 1/8 NPT