Learn More
Cole-Parmer Ashcroft 10052.5 Utility Gauge, 30 to 0" Hg, 2 1/2" Dial, Bottom Connection
Catalog No. 50992662 Shop All Cole-Parmer Products
Encompass_Preferred
Gauges include brass socket, soft-soldered phosphor bronze Bourdon tube, painted steel case, and polycarbonate lens.
Catalog No. 50-992-662 Supplier Cole-Parmer Supplier No. 6800400
Gauges include brass socket, soft-soldered phosphor bronze Bourdon tube, painted steel case, and polycarbonate lens.