Learn More
Cole-Parmer Ashcroft 1008SL2.5 30 to 0" Hg and 100 to 0 kPa Dual-Scale Gauge, Bottom Connection
Catalog No. 50992664 Shop All Cole-Parmer Products
Encompass_Preferred
Gauge construction includes acrylic lens and 316 stainless steel socket, Boudon tube, and case. Welded sealed joints protect against corrosion.
Catalog No. 50-992-664 Supplier Cole-Parmer Supplier No. 6800700
Gauge construction includes acrylic lens and 316 stainless steel socket, Boudon tube, and case. Welded sealed joints protect against corrosion.