Learn More
Ace Glass, Inc. Gauge, vacuum, McLeod, removable volumetric leg, 5mm Hg complete
SDP
Catalog No. 50979821
Encompass_Preferred
A 5MM tilting gauge which maintains a constant overflow head of mercury and maintains capillary depression of the meniscus.
Catalog No. 50-979-821 Supplier Ace Glass, Inc. Supplier No. 872805
A 5MM tilting gauge which maintains a constant overflow head of mercury and maintains capillary depression of the meniscus.