Learn More
Abcam Assay Kits, Enzyme substrates, Enzyme substrates for Luciferase,
Catalog No. 50996216
Encompass_Preferred
Assay Kits, Enzyme substrates, Enzyme substrates for Luciferase, MarkerGeneTM Gaussia Luciferase Cellular Assay Kit, Gaussia luciferase does not require ATP, catalyzes the oxidation of coelenterazine in a reaction that emits light (at 470 nm), and is secreted into the cell medium of cells grown in culture, 1 kit
Catalog No. 50-996-216 Supplier Abcam Supplier No. AB189814100TEST
Assay Kits, Enzyme substrates, Enzyme substrates for Luciferase, MarkerGeneTM Gaussia Luciferase Cellular Assay Kit, Gaussia luciferase does not require ATP, catalyzes the oxidation of coelenterazine in a reaction that emits light (at 470 nm), and is secreted into the cell medium of cells grown in culture, 1 kit