Learn More
Med Vet International McKesson USP Type VII Gauze Sponges, 8-Ply 4x4in, Non-Sterile, 200/package, 4000/case
Catalog No. 503044851
Encompass_Preferred
4" x 4" (10.2 cm x 10.2 cm) Non-Sterile 100% cotton USP Type VII Threads 19x9+/-1 Weight / 18.1g/m2 Ideal for wound dressings, wound packing and general wound care. Peel pouches and trays allow for easy opening. Not made with natural rubber latex. 200 Per Pack, 20 Packs Per Case
Catalog No. 50-304-4851 Supplier Med Vet International Supplier No. 408T
4" x 4" (10.2 cm x 10.2 cm) Non-Sterile 100% cotton USP Type VII Threads 19x9+/-1 Weight / 18.1g/m2 Ideal for wound dressings, wound packing and general wound care. Peel pouches and trays allow for easy opening. Not made with natural rubber latex. 200 Per Pack, 20 Packs Per Case