Learn More
Med Vet International Adhesive Dressing DermaRite Bordered Gauze 2 X 2 Inch Square Sterile, 00261E, Each
Catalog No. 503045078
Encompass_Preferred
This sterile wound dressing is soft, flexible and non-restrictive The first layer features a low adherent, lint-free material designed to protect the wound surface The second layer is super absorbent, removing released wound fluid The third layer is a nonwoven tape with a strong adhesive to keep the dressing secure and maintain a moist environment The 2 x 2-inch DermaRite adhesive gauze dressing is suitable for use on all skin types Square adhesive bandage comes individually wrapped to prevent contamination
Catalog No. 50-304-5078 Supplier Med Vet International Supplier No. 972459EA
This sterile wound dressing is soft, flexible and non-restrictive The first layer features a low adherent, lint-free material designed to protect the wound surface The second layer is super absorbent, removing released wound fluid The third layer is a nonwoven tape with a strong adhesive to keep the dressing secure and maintain a moist environment The 2 x 2-inch DermaRite adhesive gauze dressing is suitable for use on all skin types Square adhesive bandage comes individually wrapped to prevent contamination