Learn More
Med Vet International Adhesive Dressing DermaRite Bordered Gauze 4 X 4 Inch Square Sterile, 00262E, 25/Box
Catalog No. 503045103
Encompass_Preferred
Sterile wound dressing is soft, flexible and nonrestrictive The first layer features a low adherent, lint-free material designed to protect the wound surface The second layer is super absorbent, removing released wound fluid The third layer is a nonwoven tape with a strong adhesive to keep the dressing secure and maintain a moist environment Individually wrapped to prevent contamination Adhesive dressing is 10 x 10 centimeters with a pad of 5.5 x 6.25 centimeters
Catalog No. 50-304-5103 Supplier Med Vet International Supplier No. 867882BX
Sterile wound dressing is soft, flexible and nonrestrictive The first layer features a low adherent, lint-free material designed to protect the wound surface The second layer is super absorbent, removing released wound fluid The third layer is a nonwoven tape with a strong adhesive to keep the dressing secure and maintain a moist environment Individually wrapped to prevent contamination Adhesive dressing is 10 x 10 centimeters with a pad of 5.5 x 6.25 centimeters