Learn More
Med Vet International Nonwoven Sponge Curity™ 4 X 4 Inch, Sterile 6-Ply Square, 7084-, 2/Pack
Catalog No. 503044512
Encompass_Preferred
*Six-ply Nonwoven Rayon/polyester blend construction Cost-effective Highly absorbent Virtually lint-free 4 x 4 inches
Catalog No. 50-304-4512 Supplier Med Vet International Supplier No. 319263PK
*Six-ply Nonwoven Rayon/polyester blend construction Cost-effective Highly absorbent Virtually lint-free 4 x 4 inches