Learn More
Med Vet International Avant Gauze Non-Woven Non-Sterile Sponges, 2in X 2in, 4 Ply, 200/Bag
Catalog No. 502829529
Encompass_Preferred
Avant Gauze is Medline's standard non-woven dressing. Compared to traditional cotton gauze, Avant Gauze helps to reduce the amount of lint that is produced and is less traumatic to fragile granulating wound tissue because it's virtually non-adherent. It also has significantly faster wicking ability which promotes a healthier healing environment. Discover the perfect balance between strength and absorbency in this rayon/polyester blend gauze. Non-sterile.
Catalog No. 50-282-9529 Supplier Med Vet International Supplier No. NON25224Z
Avant Gauze is Medline's standard non-woven dressing. Compared to traditional cotton gauze, Avant Gauze helps to reduce the amount of lint that is produced and is less traumatic to fragile granulating wound tissue because it's virtually non-adherent. It also has significantly faster wicking ability which promotes a healthier healing environment. Discover the perfect balance between strength and absorbency in this rayon/polyester blend gauze. Non-sterile.