Learn More
Med Vet International Jorvet Sponge Gauze, 4"x4", 8ply, J0193C, 200/Bag
Catalog No. 5000495191
Encompass_Preferred
Our sponge gauze is 100 percent purecotton and comes in a 20 x 12 mesh. It is fullybleached and meets the USP standard.
Catalog No. 50-004-95191 Supplier Med Vet International Supplier No. JOR193C
Our sponge gauze is 100 percent purecotton and comes in a 20 x 12 mesh. It is fullybleached and meets the USP standard.