Learn More
Med Vet International Jorvet Sponge gauze, 4"x4", 12ply, J0193D, 200/Bag
Catalog No. 5000495456
Encompass_Preferred
Our sponge gauze is 100 percent pure cotton and comes in a 20 x 12 mesh. It is fully bleached and meets the USP standard.
Catalog No. 50-004-95456 Supplier Med Vet International Supplier No. JOR193D
Our sponge gauze is 100 percent pure cotton and comes in a 20 x 12 mesh. It is fully bleached and meets the USP standard.