Learn More
Dukal™ Gauze Sponges
Catalog No. 22415753
Change view
Click to view available options
Dimensions (L x W):
8 x 4 in.
Catalog No. Dimensions (L x W)
22-415-753 8 x 4 in.
1 options
This item is not returnable. View return policy
Catalog No. 22-415-753 Supplier Dukal™ Supplier No. 6842
Add to Cart
Add to Cart
This item is not returnable. View return policy
These Gauze Sponges are 100% woven cotton and ideal for a number of applications including general cleaning, dressings, prepping, packing, and debriding wounds.
  • 100% woven cotton.
  • Ideal for a number of applications including general cleaning, dressings, prepping, packing, and debriding wounds.
  • Most non-sterile sponges available in 200 or 100 per bag to suit multiple market needs.
Specifications
Sterility Sterile
Dimensions (L x W) 8 x 4 in.
Latex Free Yes
Thickness 12-Ply