Learn More
Grainger 3M, Scotch-Brite, General Purpose Hand Pad, 8 in x 4 in, Green, 10 Pads/Pack
Supplier:  Grainger 3JML1
Encompass_Preferred
The 3M 8210 N95 Particulate Respirator is a disposable respirator that helps provide reliable respiratory protection against certain non-oil based particles. It features an adjustable noseclip and nose foam to help provide a custom fit and secure seal. The respirator's cool flow exhalation valve helps reduce heat build-up inside the respirator. The collapse-resistant shell design allows the respirator to retain its shape. This N95 respirator is NIOSH-approved for at least 95% filtration efficiency against certain non-oil based particles and meets CDC guidelines for TB exposure control. It comes 20 respirators per box.
Catalog No. NC9332513