Learn More
Med Vet International Non-Woven Gauze Sponges, 2 in x 2 in 4-ply, 200/pkg
Catalog No. 502091965
Encompass_Preferred
Unique rayon/polyester formed fabric Significantly less linting than cotton gauze Virtually non-adherent More cost-effective than cotton gauze Non-sterile
Catalog No. 50-209-1965 Supplier Med Vet International Supplier No. PK224
Unique rayon/polyester formed fabric Significantly less linting than cotton gauze Virtually non-adherent More cost-effective than cotton gauze Non-sterile