Learn More
Med Vet International Caring gauze bandage, 4.5 x 4.1yd, sterile, lf , 100/Case
Catalog No. 502092018
Encompass_Preferred
Caring gauze bandage rolls are 6-ply, 100% cotton unique crimped, open-weave design means superior absorbency and cushioning. Gently stretches and conforms to body contours. Used as either a primary or secondary dressing sterile, 4.5" x 4.1 yds.
Catalog No. 50-209-2018 Supplier Med Vet International Supplier No. PRM25865
Caring gauze bandage rolls are 6-ply, 100% cotton unique crimped, open-weave design means superior absorbency and cushioning. Gently stretches and conforms to body contours. Used as either a primary or secondary dressing sterile, 4.5" x 4.1 yds.