Learn More
Med Vet International Gauze Sponges, 4x4, Non-Sterile, 200/pkg, 15 Packages
Catalog No. 502092003
Encompass_Preferred
High-quality, non-sterile gauze made of lint-free cotton with no raw edges or loose threads. High0absorbency, low linting, and costs less than the leading brands. Compare to any leading brand of medical sponge. 200 per package. Great wound care supplies for daily use in vet clinics, medical offices and hospitals These medical gauze sponges have no exposed raw edges which reduces lint Highly absorbent Non-Sterile 200 sponges per package 4"x4" 12-Ply Gauze sponges also available in 8-ply 2"x2" (PK208), 3"x3" (PK308), and 4"x4" (PK408) standard sizes.
Catalog No. 50-209-2003 Supplier Med Vet International Supplier No. PK412X15
High-quality, non-sterile gauze made of lint-free cotton with no raw edges or loose threads. High0absorbency, low linting, and costs less than the leading brands. Compare to any leading brand of medical sponge. 200 per package. Great wound care supplies for daily use in vet clinics, medical offices and hospitals These medical gauze sponges have no exposed raw edges which reduces lint Highly absorbent Non-Sterile 200 sponges per package 4"x4" 12-Ply Gauze sponges also available in 8-ply 2"x2" (PK208), 3"x3" (PK308), and 4"x4" (PK408) standard sizes.