Learn More
Med Vet International Gauze Sponges, 4x4, Non-Sterile, 8-Ply, 200/pkg
Catalog No. 502092038
Encompass_Preferred
These cotton sponges are ideal for wound dressings, wound packing and general wound care. High quality, non-sterile gauze made of lint free cotton with no raw edges or loose threads. High absorbency, low linting, and costs less than leading brands ... compare to any leading brand of medical sponge. >Great wound care supplies for daily use in veterinary clinics, medical offices and hospitals. These medical gauze sponges have no exposed raw edges that reduce lint. Highly absorbent Non-sterile 200 sponges per pkg Gauze sponges also available in 8-ply 2" x 2" (208C), 3" x 3" (308C), and 4" x 4" (408C) standard sizes.
Catalog No. 50-209-2038 Supplier Med Vet International Supplier No. PK408
These cotton sponges are ideal for wound dressings, wound packing and general wound care. High quality, non-sterile gauze made of lint free cotton with no raw edges or loose threads. High absorbency, low linting, and costs less than leading brands ... compare to any leading brand of medical sponge. >Great wound care supplies for daily use in veterinary clinics, medical offices and hospitals. These medical gauze sponges have no exposed raw edges that reduce lint. Highly absorbent Non-sterile 200 sponges per pkg Gauze sponges also available in 8-ply 2" x 2" (208C), 3" x 3" (308C), and 4" x 4" (408C) standard sizes.