Learn More
Med Vet International Kerlix Type Gauze Rolls (Non-Sterile) 3.4in x 3.1 yds., 6ply, 96/Case
Supplier:  Med Vet International BULKEE3NS
Encompass_Preferred
6-ply, 100% cotton. Unique crimped, open-weave design means superior absorbency and cushioning. Gently stretches and comforms to body contours. Used as either a primary or secondary dressing. 3.4'x 3.1 yard Kerlix-type rolls.
Catalog No. 50-209-2104