Learn More
Med Vet International Kerlix Type Gauze Rolls, 4.5in X 4.1 Yds, 6 Ply, Non-Sterile, 10/Pack
Supplier:  Med Vet International BULKEENSX10
Encompass_Preferred
6 Ply, 100% cotton. Unique crimped, open-weave design means superior absorbency and cushioning. Gently stretches and conforms to body contours. Used as either a primary or secondary dressing. 4.5' X 4.1 Yds Kerlix-type rolls.
Catalog No. 50-209-2106