Learn More
Fine Science Tools Single Use Feeding Needles, Straight, 75 mm, 5/pk
SDP
Catalog No. NC9283075
Encompass
Product: Single Use Feeding Needles - Straight/75mm/15G/3mm Tip Diameter/ Gauge: 15/ Tip Diameter: 3mm/ Shape: Straight/ Tip Shape: Barrel/ Material: Composite/ Length: 75mm/ Sterile: Yes/ Style: Single Use/ Unit
Catalog No. NC9283075 Supplier Fine Science Tools Supplier No. 1806115
Product: Single Use Feeding Needles - Straight/75mm/15G/3mm Tip Diameter/ Gauge: 15/ Tip Diameter: 3mm/ Shape: Straight/ Tip Shape: Barrel/ Material: Composite/ Length: 75mm/ Sterile: Yes/ Style: Single Use/ Unit