Learn More
Fine Science Tools Reusable Feeding Needles, 18 Gauge, 50 mm
SDP
Catalog No. NC9349775
Encompass
Reusable Feeding Needle - Curved/50mm/18G/2.25mm Tip Diameter/ Gauge: 18/Shape: Curved/ Tip Shape: Round/ Alloy: Stainless Steel/ Length: 50mm/ Sterile: No/ Style: Reusable/ Unit
Catalog No. NC9349775 Supplier Fine Science Tools Supplier No. 1806150
Reusable Feeding Needle - Curved/50mm/18G/2.25mm Tip Diameter/ Gauge: 18/Shape: Curved/ Tip Shape: Round/ Alloy: Stainless Steel/ Length: 50mm/ Sterile: No/ Style: Reusable/ Unit