Learn More
Fine Science Tools Single Use Feeding Needles, Straight, 50 mm, 5/pk
SDP
Catalog No. NC9001530
Encompass
Product: Single Use Feeding Needles - Straight/50mm/18G/2.5mm Tip Diameter/ Gauge: 18/ Tip Diameter: 2.5mm/ Shape: Straight/ Tip Shape: Barrel/ Material: Composite/ Length: 50mm/ Sterile: Yes/ Style: Single Use/ Unit
Catalog No. NC9001530 Supplier Fine Science Tools Supplier No. 1806118
Product: Single Use Feeding Needles - Straight/50mm/18G/2.5mm Tip Diameter/ Gauge: 18/ Tip Diameter: 2.5mm/ Shape: Straight/ Tip Shape: Barrel/ Material: Composite/ Length: 50mm/ Sterile: Yes/ Style: Single Use/ Unit