Learn More
TARGETMOL CHEMICALS INC GAVESTINEL 5MG
SDP
Catalog No. 502580921
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Gavestinel (GV 150526) is a non-competitive NMDA receptor antagonist that is potent selective and orally active.Gavestinel binds to the glycine site of the NMDA receptor with a binding affinity (pKi value) of 8.5.Gavestinel is used in acute ischaemic stroke studies. purity: 99%
Catalog No. 50-258-0921 Supplier TARGETMOL CHEMICALS INC Supplier No. T227985MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Gavestinel (GV 150526) is a non-competitive NMDA receptor antagonist that is potent selective and orally active.Gavestinel binds to the glycine site of the NMDA receptor with a binding affinity (pKi value) of 8.5.Gavestinel is used in acute ischaemic stroke studies. purity: 99%