Learn More
Thomas Scientific Pycnometer, 25 ml (+ / -0.05)
SDP
Catalog No. NC0829086
Encompass_Preferred
Gay-Lussac With capillary glass stopper ground to flat-sided point for take-off of overflow 10 mL and 25 mL sizes are for ASTM Standard D 369 and Barrett test H-8; AASHTO standard T-74
Catalog No. NC0829086 Supplier Thomas Scientific Supplier No. 8350B40
Gay-Lussac With capillary glass stopper ground to flat-sided point for take-off of overflow 10 mL and 25 mL sizes are for ASTM Standard D 369 and Barrett test H-8; AASHTO standard T-74