Learn More
Braintree Scientific Heated Hard Pad Connectors for Gaymar Pump
SDP
Catalog No. NC0227300
Encompass
Water Heated Hard Pads - Connects to Gaymar, Androit and other pumps. Durable polycarbonate construction. Scalpel, needle, and towel clamp proof. Smooth surfaces are easy to clean helping to prevent nosocomial infection. Highly puncture resistant. **Sold for research purposes only and not for human use**
Catalog No. NC0227300 Supplier Braintree Scientific Supplier No. HHPGAYMAR
Water Heated Hard Pads - Connects to Gaymar, Androit and other pumps. Durable polycarbonate construction. Scalpel, needle, and towel clamp proof. Smooth surfaces are easy to clean helping to prevent nosocomial infection. Highly puncture resistant. **Sold for research purposes only and not for human use**