Learn More
Braintree Scientific Gaymar Multi-T Pad - Single Patient Use,18 x 26, 10/case
SDP
Catalog No. 501956060
Encompass
Gaymar produces a wide variety of MulTiPad products for use with the T/Pump. The random flow design permits the pad to be folded to almost any size, reducing the need for added inventory. Together, the T/Pump and MulTiPad deliver a consistent flow of regulated water temperature for safe, effective warming and cooling. Clik-Tite connectors are leak-proof and multiple pads can be connected to one unit to surround your patient with continuous warmth. Unique flow design provides optimum therapy even when folded. Pads can be folded to almost any size, reducing the number of pads your facility needs in inventory. Single Patient Use - Material: Pliable polymer/non-women fabric on one side. **Sold for research purposes only and not for human use**
Catalog No. 50-195-6060 Supplier Braintree Scientific Supplier No. TPD26E
Gaymar produces a wide variety of MulTiPad products for use with the T/Pump. The random flow design permits the pad to be folded to almost any size, reducing the need for added inventory. Together, the T/Pump and MulTiPad deliver a consistent flow of regulated water temperature for safe, effective warming and cooling. Clik-Tite connectors are leak-proof and multiple pads can be connected to one unit to surround your patient with continuous warmth. Unique flow design provides optimum therapy even when folded. Pads can be folded to almost any size, reducing the number of pads your facility needs in inventory. Single Patient Use - Material: Pliable polymer/non-women fabric on one side. **Sold for research purposes only and not for human use**